Xanthine ≥99%
100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 69-89-6 Molecular Formula: C5H4N4O2 Formula Weight: 152.11 Odour: Odourless Melting Point: 300° Beilstein Reference: 8733 Merck Reference: 14,10059 MDL No.: MFCD00078453
Synonyms: 1H-Purine-2,6(3H,7H)-dione, 3,7-Dihydro-1H-purine-2,6-dione, 2,6-Dihydroxypurine
- CAS Number:69-89-6
- Formula:C₅H₄N₄O₂
- MDL Number:MFCD00078453
- Molecular Weight:152.11 g/mol
- Merck Index:14,10059
- Melting Pt:>300 °C
- Storage Temperature:Ambient
- EINECS:200-718-6
Documents
Specifications
Specifications
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



