Order Entry
Germany
Orders LinkContactUsLinkComponent
Ninhydrin ≥99%

Ninhydrin ≥99%

Glass bottle for solids, 250 g
Catalog # A10409.30
Reizend
Ninhydrin ≥99%
Catalog # A10409.30
CAS Number:  485-47-2
Ninhydrin ≥99%
Ninhydrin ≥99%

Glass bottle for solids, 250 g

Wechseln zu:
CAS Number:  485-47-2
Price
365,00 €
Anmelden für die Verfügbarkeit
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 485-47-2 Molecular Formula: C9H6O4 Formula Weight: 178.14 Melting Point: 251° dec. Beilstein Reference: 1910963 Merck Reference: 14,6554 MDL No.: MFCD00003791

Synonyme:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS-Nummer:485-47-2
  • Formel::C₉H₆O₄
  • MDL-Nummer:MFCD00003791
  • Molekulargewicht:178.14 g/mol
  • Merck-Index:14,6554
  • Siedepunkt:351 °C (1013 hPa)
  • Schmelzpunkt:251 °C (dec.)
  • Dichte:0.862 g/cm³ (20 °C)
  • Lagertemperatur:Ambient
  • EINECS:207-618-1
Spezifikationen:

Spezifikationen:

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Spezifikationen:

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.