Order Entry
Germany
Orders LinkContactUsLinkComponent
Ninhydrin ≥99%

Ninhydrin ≥99%

Glass bottle for solids, 10 g
Catalog # A10409.09
Irritant
Ninhydrin ≥99%
Catalog # A10409.09
CAS Number:  485-47-2
Ninhydrin ≥99%
Ninhydrin ≥99%

Glass bottle for solids, 10 g

CAS Number:  485-47-2
Price
30,80 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 99% CAS No.: 485-47-2 Molecular Formula: C9H6O4 Formula Weight: 178.14 Melting Point: 251° dec. Beilstein Reference: 1910963 Merck Reference: 14,6554 MDL No.: MFCD00003791

Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Formula:C₉H₆O₄
  • MDL Number:MFCD00003791
  • Molecular Weight:178.14 g/mol
  • Merck Index:14,6554
  • Boiling Pt:351 °C (1013 hPa)
  • Melting Pt:251 °C (dec.)
  • Density:0.862 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-618-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications