Order Entry
Germany
Orders LinkContactUsLinkComponent
 
The site is currently experiencing technical difficulties. Please retry later. We apologize for any inconvenience.

Pyrocatechol ≥99%

Glass bottle for solids, 250 g
Catalog # A10164.30
Toxic
Corrosive
Health
Pyrocatechol ≥99%
Catalog # A10164.30
CAS Number:  120-80-9
Pyrocatechol ≥99%
Pyrocatechol ≥99%

Glass bottle for solids, 250 g

CAS Number:  120-80-9
Price
35,80 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 120-80-9 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 127°(260°F) Physical Form: Powder or flakes Density: 1.370 Sensitivity: Air and Light Sensitive Appearance: White to brown Odour: Aromatic Melting Point: 104-107° Boiling Point: 245-246° Beilstein Reference: 471401 Merck Reference: 14,7999 UN No.: 2811 MDL No.: MFCD00002188

Synonyms:  o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
  • CAS Number:120-80-9
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002188
  • Molecular Weight:110.11 g/mol
  • Merck Index:14,7999
  • Boiling Pt:245…246 °C
  • Melting Pt:104…107 °C
  • Density:1.370
  • Flash Pt:127 °C (260 °F)
  • Storage Temperature:Ambient
  • EINECS:204-427-5
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications