Pyrocatechol ≥99%
Glass bottle for solids, 1.000 g
Hersteller: Thermo Fisher Scientific
Wechseln zu:
Product Details & Documents
Purity: 99% CAS No.: 120-80-9 Molecular Formula: C6H6O2 Formula Weight: 110.11 Flash Point: 127°(260°F) Physical Form: Powder or flakes Density: 1.370 Sensitivity: Air and Light Sensitive Appearance: White to brown Odour: Aromatic Melting Point: 104-107° Boiling Point: 245-246° Beilstein Reference: 471401 Merck Reference: 14,7999 UN No.: 2811 MDL No.: MFCD00002188
Synonyme: o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
- CAS-Nummer:120-80-9
- Formel::C₆H₆O₂
- MDL-Nummer:MFCD00002188
- Molekulargewicht:110.11 g/mol
- Merck-Index:14,7999
- Siedepunkt:245…246 °C
- Schmelzpunkt:104…107 °C
- Dichte:1.370
- Flammpunkt:127 °C (260 °F)
- Lagertemperatur:Ambient
- EINECS:204-427-5
- UN:2811
- ADR:6.1,III
Dokumente
Zertifikate
Suche nach Chargennummermsds_ALFAA10164
Sicherheitsdatenblatt
Organic Building Blocks and Intermediates
Anwendungshinweise
Spezifikationen:
Spezifikationen:
- Pack type:Glass bottle for solids
- Pk:1.000 g
Spezifikationen:
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



