Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Polysorbate 80

Glass bottle, 2.500 mL
Catalog # L13315.0F
Polysorbate 80
Catalog # L13315.0F
CAS Number:  9005-65-6
Polysorbate 80
Polysorbate 80

Glass bottle, 2.500 mL

CAS Number:  9005-65-6
Price
158,00 €
Log in for availability
1 * 2.500 mL
Product Details & Documents

CAS No.: 9005-65-6 Flash Point: 149°(300°F) Refractive Index: 1.4750 Boiling Point: 100° Merck Reference: 14,7582 MDL No.: MFCD00082107

Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Merck Index:14,7582
  • Boiling Pt:>100 °C
  • Melting Pt:100 °C
  • Density:1.064
  • Flash Pt:>149 °C (300 °F)
  • Storage Temperature:Ambient
  • EINECS:500-019-9
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications