Order Entry
Germany
Orders LinkContactUsLinkComponent
Copper(II) oxide, nanopowder

Copper(II) oxide, nanopowder

50 g
Catalog # 44663.18
Irritant
Environment
Copper(II) oxide, nanopowder
Catalog # 44663.18
CAS Number:  1317-38-0
Copper(II) oxide, nanopowder
Copper(II) oxide, nanopowder

50 g

CAS Number:  1317-38-0
Price
149,00 €
Log in for availability
1 * 50 g
Product Details & Documents

CAS No.: 1317-38-0 Molecular Formula: CuO Formula Weight: 79.55 Physical Form: 30-50nm APS Powder, S.A. 13m2/g Density: 6.3 Refractive Index: 2.63 Odour: Odourless Melting Point: 1326° Merck Reference: 14,2646 UN No.: 3077 MDL No.: MFCD00010979

Synonyms:  Cupric oxide, Copper oxide, Copper monoxide granulated
  • CAS Number:1317-38-0
  • Formula:CuO
  • MDL Number:MFCD00010979
  • Molecular Weight:79.55 g/mol
  • Merck Index:14,2646
  • Boiling Pt:2000 °C (1013 hPa)
  • Melting Pt:1326 °C
  • Density:6.3
  • Storage Temperature:Ambient
  • EINECS:215-269-1
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.