Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Copper(II) oxide, nanopowder

10 g
Catalog # 44663.09
Irritant
Environment
Copper(II) oxide, nanopowder
Catalog # 44663.09
CAS Number:  1317-38-0
Copper(II) oxide, nanopowder
Copper(II) oxide, nanopowder

10 g

CAS Number:  1317-38-0
Price
45,20 €
Log in for availability
1 * 10 g
Product Details & Documents

CAS No.: 1317-38-0 Molecular Formula: CuO Formula Weight: 79.55 Physical Form: 30-50nm APS Powder, S.A. 13m2/g Density: 6.3 Refractive Index: 2.63 Odour: Odourless Melting Point: 1326° Merck Reference: 14,2646 UN No.: 3077 MDL No.: MFCD00010979

Synonyms:  Cupric oxide, Copper oxide, Copper monoxide granulated
  • CAS Number:1317-38-0
  • Formula:CuO
  • MDL Number:MFCD00010979
  • Molecular Weight:79.55 g/mol
  • Merck Index:14,2646
  • Boiling Pt:2000 °C (1013 hPa)
  • Melting Pt:1326 °C
  • Density:6.3
  • Storage Temperature:Ambient
  • EINECS:215-269-1
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pk:
    10 g

Specifications