Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Viscosity Reference sample S60 1 * 500 mL
Catalog # PSLE2700-V09
Supplier:
POULTEN SELFE LEE
Expand image
Viscosity Reference sample S60 1 * 500 mL
Catalog # PSLE2700-V09
Supplier:
POULTEN SELFE LEE