ContactUsLinkComponent
Flask volumetric, capacity 100ml, class A, amber graduated, polystoppered, tolerance 0.1ml, height 170mm, NS-ground 14/23, graduatio n color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264018108
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 100ml, class A, amber graduated, polystoppered, tolerance 0.1ml, height 170mm, NS-ground 14/23, graduatio n color white, conformity certified,DIN/ISO product with single certificate. 1 * 1 items
Catalog # HIRS264018108
Supplier:  HIRSCHMANN
Frequently Bought Together