Shaking incubators, multitron standard
Product Details & Documents
For applications in microbiology and cell culture, bacteria, yeasts and fungi that require vigorous agitation for optimal results. Also ideal for cell culture where slow and gentle agitation is required or molecular biology for the preparation of Mini and Maxi- preps or for the expression of recombinant proteins. Thanks to the front opening door which supports the tray, the Multitron Standard is simple to use. It provides easy and secure access to all the flasks, regardless of their positions on the tray. The extracted tray is about 1,30 meters above the ground. The lower unit will not need you to dive inside the chamber to manipulate your samples; Simply pull the tray on the door.
- Three basic configurations where different options of refrigeration and orbit (25 and 50 mm) are available
- Accurate temperature control and two large LEDs for a visual inspection of the device status
- Control all units in a stacked group from any control panel
- Standard Multitron is available with refrigeration, a cooling loop is installed in each unit in the ventilation circuit. The main advantage of this compared to the Peltier technology systems is better cooling performance and lower energy consumption
- The front door opens downwards and can be used as a mounting for the shaker table
Ordering information: Supplied without tray. Universal tray adhesive or completed by clamps, pre- fitted trays pliers, special trays for microplates, stainless steel or plastic clamps, tube racks or microplates are available, please contact us.
Specifications
- Weight:405 kg
- Description:Triple Multitron Standard with cooling (amplitude 25 mm)
- W×D×H:1070×860×1990 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Steel flask clamp, 100 ml
Product Information
- Catalog #AvailabilityPrice
- Steel flask clamp, 1000 ml
Product Information
- Catalog #AvailabilityPrice
- Steel flask clamp, 2000 ml
Product Information
- Catalog #AvailabilityPrice
- Steel flask clamp, 25 ml
Product Information
- Catalog #AvailabilityPrice
- Steel flask clamp, 250 ml
Product Information
- Catalog #AvailabilityPrice
- Steel flask clamp, 50 ml
Product Information
- Catalog #AvailabilityPrice
- Steel flask clamp, 500 ml
Product Information
- Catalog #AvailabilityPrice
- Steel flask clamp, 5000 ml
Product Information
- Catalog #AvailabilityPrice
- Steel flask clamp, 4000 ml
Product Information
- Catalog #AvailabilityPrice
- Steel flask clamp, 3000 ml
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



