Order Entry
Germany
Orders LinkContactUsLinkComponent
TRIS BUFFER, 50 MM SOLUTION

TRIS BUFFER, 50 MM SOLUTION

Catalog # CABRS50-642
Marketsource
Supplier:   Lonza
TRIS BUFFER, 50 MM SOLUTION
Catalog # CABRS50-642
Supplier:  Lonza
Marketsource
TRIS BUFFER, 50 MM SOLUTION
Price
81,20 €
Log in for availability
1 * 30 mL
Product Details & Documents

Documents

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.