Transfer pipettes
6,0 ml, 20 - 30 µl, 146 mm, - ml, +, 10 per bag
SARS86.1175.010
Marketsource
: SARSTEDT
LDPE, disposable.
- No risk of injury or infection from broken glass
- Disposable: No carry over or cross contamination
- Can be heat-sealed for sample storage and transport
- Environmentally neutral, ensuring non-toxic disposal
- Packed:10 per bag
- Sterile:+
- Capacity:6,0 ml
- Length:146 mm
- Drop vol.:20 - 30 µl
- Graduation:- ml



