Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Pentylcyclohexanone ≥98%

4-Pentylcyclohexanone ≥98%

Glass bottle for solids, 10 g
Catalog # B22088.09
4-Pentylcyclohexanone ≥98%
Catalog # B22088.09
CAS Number:  61203-83-6
4-Pentylcyclohexanone ≥98%
4-Pentylcyclohexanone ≥98%

Glass bottle for solids, 10 g

CAS Number:  61203-83-6
Price
48,80 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 98+% CAS No.: 61203-83-6 Molecular Formula: C11H20O Formula Weight: 168.28 UN No.: 3082 MDL No.: MFCD00059581

  • CAS Number:61203-83-6
  • Formula:C₁₁H₂₀O
  • MDL Number:MFCD00059581
  • Molecular Weight:168.28 g/mol
  • Density:0.89
  • Storage Temperature:Ambient
  • EINECS:406-670-4
  • UN:3082
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications