Order Entry
Germany
Orders LinkContactUsLinkComponent
 

N,N-Dimethylformamide

5 L
Catalog # KRAE109937
Flammable
Health
Irritant
Supplier:   KRAEMER MARTIN
N,N-Dimethylformamide
Catalog # KRAE109937
Supplier:  KRAEMER MARTIN
CAS Number:  68-12-2
N,N-Dimethylformamide
N,N-Dimethylformamide

5 L

CAS Number:  68-12-2
Price
350,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09 g/mol
  • Merck Index:13,03269
  • Boiling Pt:153 °C (1013 hPa)
  • Melting Pt:–61 °C
  • Density:0.949 g/cm³ (20 °C)
  • Flash Pt:57.5 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-679-5
  • UN:2265
  • ADR:3,III
  • REACH:01-2119475605-32
Specifications

Specifications

  • Pk:
    5 L

Specifications