N,N-Dimethylformamide
Glass bottle, 1 L
KRAE24-030.1000
: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
- :68-12-2
- :HCON(CH₃)₂
- :MFCD00003284
- :73.09 g/mol
- :13,03269
- :153 °C (1013 hPa)
- :–61 °C
- :0.949 g/cm³ (20 °C)
- :57.5 °C (closed cup)
- :Ambient
- :200-679-5
- :2265
- :3,III
- :01-2119475605-32
- Pack type:Glass bottle
- Pk:1 L