Order Entry
Germany
Orders LinkContactUsLinkComponent
Ethyl acetate

Ethyl acetate

Plastic bottle, 5 L
Catalog # KRAE17-150.5000
Flammable
Irritant
Supplier:   KRAEMER MARTIN
Ethyl acetate
Catalog # KRAE17-150.5000
Supplier:  KRAEMER MARTIN
CAS Number:  141-78-6
Ethyl acetate
Ethyl acetate

Plastic bottle, 5 L

CAS Number:  141-78-6
Price
61,80 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Acetic acid ethyl ester
  • CAS Number:141-78-6
  • Formula:CH₃COOC₂H₅
  • MDL Number:MFCD00009171
  • Molecular Weight:88.11 g/mol
  • Merck Index:13,03792
  • Boiling Pt:77.1 °C (1013 hPa)
  • Melting Pt:–83 °C
  • Density:0.902 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • EINECS:205-500-4
  • UN:1173
  • ADR:3,II
  • REACH:01-2119475103-46

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.