Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Ethyl acetate

Glass bottle, 1 L
Catalog # KRAE17-150.1000
Flammable
Irritant
Supplier:   KRAEMER MARTIN
Ethyl acetate
Catalog # KRAE17-150.1000
Supplier:  KRAEMER MARTIN
CAS Number:  141-78-6
Ethyl acetate
Ethyl acetate

Glass bottle, 1 L

CAS Number:  141-78-6
Price
34,10 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Acetic acid ethyl ester
  • CAS Number:141-78-6
  • Formula:CH₃COOC₂H₅
  • MDL Number:MFCD00009171
  • Molecular Weight:88.11 g/mol
  • Merck Index:13,03792
  • Boiling Pt:77.1 °C (1013 hPa)
  • Melting Pt:–83 °C
  • Density:0.902 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • EINECS:205-500-4
  • UN:1173
  • ADR:3,II
  • REACH:01-2119475103-46

 

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications