Basic cleanroom benches
Basic cleanroom bench, 600×350×450 mm
Catalog # 134-0496
Supplier: Palbam
Jump to:
Product Details & Documents
Cleanroom benches in stainless steel 304 with electropolished finish.
- Rounded edges
- Height adjustable legs
- Leg covers (covers unsightly bolt connections to floor)
- No snag points
- Ultra-clean
Delivery information: Typically available from stock for direct shipment from the supplier.
Documents
Palbam Class - Catalog 2018
Product Literature
Specifications
- Description:Basic cleanroom bench
- W×D×H:600×350×450 mm