ContactUsLinkComponent
Flask volumetric, capacity 250ml, with rim, DURAN, class A, tolerance 0.15ml, height 220mm, graduation color blue, conformity certif ied. 1 * 1 items
Catalog # HIRS2800186_S
Supplier:  HIRSCHMANN
undefined
Flask volumetric, capacity 250ml, with rim, DURAN, class A, tolerance 0.15ml, height 220mm, graduation color blue, conformity certif ied. 1 * 1 items
Catalog # HIRS2800186_S
Supplier:  HIRSCHMANN
Frequently Bought Together