Orders LinkContactUsLinkComponent
Pressure Bulbs
Pressure Bulbs
Atomiser bulb with suction valve, red-brown, size 3, PVC, 65
Catalog # DENE4250003
Supplier:   DEUTSCH NEUMANN
Pressure Bulbs
Catalog # DENE4250003
Supplier:  DEUTSCH NEUMANN
Pressure Bulbs
Pressure Bulbs
Atomiser bulb with suction valve, red-brown, size 3, PVC, 65
Price
34,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Specifications
  • Capacity:
    65
  • Description:
    Atomiser bulb with suction valve, red-brown, size 3, PVC
  • Seq:
    2