Temperature indicating tubes
For autoclaves up to 125 °C
Catalog # KOEHK-1250
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Jump to:
Product Details & Documents
Temperature indicating tubes are used for autoclaves and hot air sterilisers.
Sterilisation is detected by the colour change of the control tube from red to green. The tubes should be placed as close as possible next to or in the load.
Specifications
- Description:For autoclaves up to 125 °C
- Seq:01