Order Entry
Germany
Orders LinkContactUsLinkComponent
Accessories for fluid and solid samplers

Accessories for fluid and solid samplers

Cleaning brush, PVC/stainless steel, Ø: 50 mm, 1000 mm
Catalog # 300-0110
Supplier:   BURKLE
Accessories for fluid and solid samplers
Catalog # 300-0110
Supplier:  BURKLE
Accessories for fluid and solid samplers
Accessories for fluid and solid samplers

Cleaning brush, PVC/stainless steel, Ø: 50 mm, 1000 mm

Price
59,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Cleaning brush, PVC/stainless steel, Ø: 50 mm, 1000 mm
  • Seq:
    5
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.