Orders LinkContactUsLinkComponent
Screw caps
GL 56
Catalog # VITL83317
Supplier:   VITLAB
Screw caps
Catalog # VITL83317
Supplier:  VITLAB
Screw caps
Screw caps
GL 56
Price
36,00 €
Log in for availability
1 * 12 ST
Product Details & Documents
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
Specifications
  • Thread:
    GL 56