Orders LinkContactUsLinkComponent
Screw caps
GL 52
Catalog # VITL83316
Supplier:   VITLAB
Screw caps
Catalog # VITL83316
Supplier:  VITLAB
Screw caps
Screw caps
GL 52
Price
32,70 €
Log in for availability
1 * 12 ST
Product Details & Documents
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
Specifications
  • Thread:
    GL 52