Orders LinkContactUsLinkComponent
Temperature indicating tubes
Temperature indicating tubes
For hot-air sterilization 180 °C/12 min
Catalog # 391-0085
Temperature indicating tubes
Catalog # 391-0085
Temperature indicating tubes
Temperature indicating tubes
For hot-air sterilization 180 °C/12 min
Price
161,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Temperature indicating tubes are used for autoclaves and hot air sterilisers.
Sterilisation is detected by the colour change of the control tube from red to green. The tubes should be placed as close as possible next to or in the load.
Specifications
  • Description:
    For hot-air sterilization 180 °C/12 min
  • Seq:
    02