Orders LinkContactUsLinkComponent
Safety shields
Safety shields
Straight, 900×10×670 mm
Catalog # 113-5031
Safety shields
Catalog # 113-5031
Safety shields
Safety shields
Straight, 900×10×670 mm
Price
475,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Protective shield made from acrylic glass to be used as splinter shield and splash guard.
  • Strong and stable design
  • Removable feet
Specifications
  • Form:
    Straight
  • W×D×H:
    900×10×670 mm