Order Entry
Germany
Orders LinkContactUsLinkComponent
Test gas cans

Test gas cans

Test gas, 100 ppm isobutylene, capacity 34 l
Catalog # 111-1563
Supplier:   M S A
Test gas cans
Catalog # 111-1563
Supplier:  M S A
Test gas cans
Test gas cans

Test gas, 100 ppm isobutylene, capacity 34 l

Price
212,76 €
Log in for availability
1 * 1 ST
Product Details & Documents

These test gases are suitable for use with wearable gas devices.

Specifications
  • Description:
    Test gas, 100 ppm isobutylene, capacity 34 l
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.