ContactUsLinkComponent
Bromide bromate solution 0.01235 mol Br2/l = 1/243 mol KBrO3/l 0.0247 N solution Content: 2,5 l 1 * 2.500 mL
Catalog # KRAF10933.4700
Supplier:  Bernd Kraft
Bromide bromate solution 0.01235 mol Br2/l = 1/243 mol KBrO3/l 0.0247 N solution Content: 2,5 l 1 * 2.500 mL
Catalog # KRAF10933.4700
Supplier:  Bernd Kraft