Order Entry
Germany
Orders LinkContactUsLinkComponent
Diethyl ether stabilised, extra pure

Diethyl ether stabilised, extra pure

25 L
Catalog # KRAF07106.8900
Flammable
Irritant
Supplier:   Bernd Kraft
Diethyl ether stabilised, extra pure
Catalog # KRAF07106.8900
Supplier:  Bernd Kraft
CAS Number:  60-29-7
Diethyl ether stabilised, extra pure
Diethyl ether stabilised, extra pure

25 L

CAS Number:  60-29-7
Price
553,00 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 7 ppm
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Specifications

Specifications

  • Pk:
    25 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.