Diethyl ether stabilised, extra pure
5 L
KRAF07106.5900
: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- :60-29-7
- :(CH₃CH₂)₂O
- :MFCD00011646
- :74.12 g/mol
- :12,03852
- :35 °C (1013 hPa)
- :–123 °C
- :0.7135 g/cm³ (20 °C)
- :–40 °C
- :1155
- :3,I
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 7 ppm
- :Ambient
- :200-467-2
- :01-2119535785-29
- Pk:5 L
- Cat. No.:KRAF07106.5900