Pyridin, reinst
Glass bottle, 2.500 mL
Catalog # KRAF07051.4700
Supplier: Bernd Kraft
Jump to:
Product Details & Documents
Synonyms: Azabenzene, Azine
- CAS Number:110-86-1
- Formula:C₅H₅N
- MDL Number:MFCD00011732
- Molecular Weight:79.1 g/mol
- Merck Index:13,08060
- Boiling Pt:115 °C (1013 hPa)
- Melting Pt:–44 °C
- Density:0,983 g/cm³ (20 °C)
- Flash Pt:17 °C
- Storage Temperature:Raumtemperatur
- EINECS:203-809-9
- UN:1282
- ADR:3,II
Specifications
Specifications
- Packung:Glass bottle
- VE:2.500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



