Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyridine, extra pure

Pyridine, extra pure

Glass bottle, 1 L
Artikelnummer KRAF07051.3700
Flammable
Irritant
Hersteller:   Bernd Kraft
Pyridine, extra pure
Artikelnummer KRAF07051.3700
Hersteller:  Bernd Kraft
CAS-Nummer:  110-86-1
Pyridine, extra pure
Pyridine, extra pure

Glass bottle, 1 L

CAS-Nummer:  110-86-1
Preis
124,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Azabenzene, Azine
  • CAS Number:110-86-1
  • Formula:C₅H₅N
  • MDL Number:MFCD00011732
  • Molecular Weight:79.1 g/mol
  • Merck Index:13,08060
  • Boiling Pt:115 °C (1013 hPa)
  • Melting Pt:–44 °C
  • Density:0.983 g/cm³ (20 °C)
  • Flash Pt:17 °C
  • Storage Temperature:Ambient
  • EINECS:203-809-9
  • UN:1282
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.