L(+)-Ascorbic acid, extra pure
Plastic bottle for solids, 500 g
Catalog # KRAF06134.2600
Supplier: Bernd Kraft
Jump to:
Product Details & Documents
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
- Merck Index:13,00837
- Boiling Pt:553 °C (1013 hPa)
- Melting Pt:190…192 °C
- Density:1.694 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-066-2
- REACH:exempt
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:500 g



