Nitric acid approx. 1 mol/l - approx. 1 M solution contains 0.2 - 0.5 % polyvinyl alcohol Content: 10 l 1 * 10 L
Catalog # KRAF05912.6020
Supplier: Bernd Kraft
Nitric acid approx. 1 mol/l - approx. 1 M solution contains 0.2 - 0.5 % polyvinyl alcohol Content: 10 l 1 * 10 L
Catalog # KRAF05912.6020
Supplier: Bernd Kraft
Frequently Bought Together
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Toluene 99.5% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylacetamide 99+%, extra pure
Supplier: Thermo Fisher Scientific
Dichloromethane 99.8% stabilised for HPLC
Supplier: Thermo Fisher Scientific
1-Propanol ACS for spectroscopy
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Water with 0,1% (v/v) acetic acid, CHROMASOLV™ LC-MS LabReady™, Solvent Blend, Riedel-de Haën™
Supplier: Honeywell Chemicals
Cyclohexane 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Toluene ≥99.7% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Xylene (mixture of isomers), J.T.Baker®
Supplier: Avantor
Methanol ≥99.8% for HPLC
Supplier: VWR Chemicals
Chloroform 99.8% stabilised for spectroscopy
Supplier: Thermo Fisher Scientific
tert-Butyl methyl ether ≥99.8%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Hexanol 98%, Reagent Grade
Supplier: Honeywell Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND