Ammonium iron(III) sulfate solution 120 g/l pure 120 g (NH4)Fe(SO4)2 * 12 H2O + 623 ml HNO3 65 %/l in diluted nitric acid Content: 1 l 1 * 1 L
Catalog # KRAF05578.3000
Supplier: Bernd Kraft
Ammonium iron(III) sulfate solution 120 g/l pure 120 g (NH4)Fe(SO4)2 * 12 H2O + 623 ml HNO3 65 %/l in diluted nitric acid Content: 1 l 1 * 1 L
Catalog # KRAF05578.3000
Supplier: Bernd Kraft
Frequently Bought Together
Disinfectant wipes, Meliseptol® HBV Tissues
Supplier: B. Braun
n-Pentane ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Methanol ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Chloroform ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Carbon disulphide ≥99.9%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS for analysis
Supplier: VWR Chemicals
VWR® IPASEPT® 70 Cleaning Solution
Supplier: VWR Chemicals
Ethyl acetate, EMPLURA®, Supelco®
Supplier: Merck
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Supplier: VWR Chemicals
n-Heptane ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Mercury thiocyanate solution 3 g Hg(SCN)2/l in methanol Content: 1 l 1 * 1 L
Supplier: Bernd Kraft
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Hellmanex III Spec. Cleaning f. cuvette 1 * 1 L
Supplier: Merck