ICP standard zinc 1.000 g Zn/l Zn in nitric acid 0.5 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Catalog # KRAF03849.0000
Supplier: Bernd Kraft
ICP standard zinc 1.000 g Zn/l Zn in nitric acid 0.5 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Catalog # KRAF03849.0000
Supplier: Bernd Kraft
Frequently Bought Together
UPC2 QC REFERENCE MATERIAL 1 * 1 KIT
Supplier: WATERS
GlycoWorks Phosphoglycan SPE Reagents HILIC 1 * 1 KIT
Supplier: WATERS
Calcium Standard for AAS, TraceCERT TM 1 * 250 mL
Supplier: Merck
Nitrate standards for ISE calibration, Orion™
Supplier: Thermo Orion
Supelco® Organic Reference Standard, α-Tocotrienol
Supplier: Merck
in Acetonitrile 1 * 1 mL
Supplier: EHRENSTORFER
Pymetrozine certified 1 * 0,1 g
Supplier: EHRENSTORFER
Nitenpyram Analytical standard, reference material, Fluka™
Supplier: Honeywell Chemicals
Pesticide reference standard, Endrin 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Supplier: EHRENSTORFER
Supelco® Organic Reference Standard, α-Arbutin
Supplier: Merck
Formoterol Related Compounds, Reference Standard
Supplier: USP
Picrocrocin phyproof(R) Reference Substance 1 * 10 mg
Supplier: Merck
Supelco® TraceCERT® Organic Reference Standard, Dodin
Supplier: Merck