ICP standard selenium 1.000 g Se/l SeO2 in nitric acid 0.5 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
KRAF03802.0000
ICP standard selenium 1.000 g Se/l SeO2 in nitric acid 0.5 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
KRAF03802.0000
Frequently Bought Together
CEFAZOLIN SODIUM SALT 1 * 75 mg
: Merck
DEXTRAN CALIBRATION LADDER
: WATERS
Terpene Mixture 2500µg/mL in Hexane 1 * 1 mL
: EHRENSTORFER
BREVETOXIN 3 SOLUTION 1 * 0,5 mL
: Merck
n-Pentadecane, Supelco®
: Merck
n-Octane, Supelco®
: Merck
VWR®, Flow Cup viscosity standards
: VWR Chemicals