0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
ICP standard tellurium 1.000 g Te/l Te in nitric acid 3 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
KRAF03789.0000
:
Bernd Kraft
ICP standard tellurium 1.000 g Te/l Te in nitric acid 3 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
KRAF03789.0000
:
Bernd Kraft
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
: Merck
Buffer solutions in sachets, Certipur®, Supelco®
: Merck
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, DuraCal
: HAMILTON BONADUZ
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
pH buffers, ready to use
: WTW
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
: VWR Chemicals
pH buffer, single-use, SINGLET™
: Hach
Cesium standard solution, 1000 mg/L Cs in 0,5 M hydrochloric acid (from Cs₂CO₃) standard for ICP
: Bernd Kraft