0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
ICP standard vanadium 1.000 g V/l V2O5 in nitric acid 1 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
KRAF03784.0000
:
Bernd Kraft
ICP standard vanadium 1.000 g V/l V2O5 in nitric acid 1 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
KRAF03784.0000
:
Bernd Kraft
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
: Merck
pH buffers, DuraCal
: HAMILTON BONADUZ
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
: Merck
pH buffers, ready to use
: WTW
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
: VWR Chemicals