ZrOCl₂ in Salzsaure 2 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF03745.0000
Supplier: Bernd Kraft
ZrOCl₂ in Salzsaure 2 mol/l ruckfuhrbar auf NIST 1 * 100 mL
Catalog # KRAF03745.0000
Supplier: Bernd Kraft
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
epT.I.P.S.® Reloads, Pipette Tips
Supplier: EPPENDORF
UV/Visible standards, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ