Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer solutions, ready to use

4,66, Acetate, Colourless, 20 °C, Plastic bottle, 1 L
Catalog # KRAF03397.3000
Supplier:   Bernd Kraft
Buffer solutions, ready to use
Catalog # KRAF03397.3000
Supplier:  Bernd Kraft
CAS Number:  
Buffer solutions, ready to use
Buffer solutions, ready to use

4,66, Acetate, Colourless, 20 °C, Plastic bottle, 1 L

CAS Number:  
Price
39,80 €
Log in for availability
1 * 1 L
Product Details & Documents
  • NIST traceable

 

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Acetate
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • Colour:
    Colourless
  • pH value:
    4,66

Specifications