Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer solution, colour coded, ready to use

9,0, Borate, Blue, 20 °C, Plastic bottle, 1 L
  KRAF03286.3060
Buffer solution, colour coded, ready to use
  KRAF03286.3060
 :  
Buffer solution, colour coded, ready to use
Buffer solution, colour coded, ready to use

9,0, Borate, Blue, 20 °C, Plastic bottle, 1 L

 :  
 
45,20 €
   
1 * 1 L
 
  • NIST traceable
  • Colour coded with choice of colours for pH values 4,00 and 7,00
 

 

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Borate
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • Colour:
    Blue
  • pH value:
    9,0

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.