Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer solutions, ready to use

8,0, Phosphate, Colourless, 20 °C, Plastic bottle, 1 L
  KRAF03087.3060
Buffer solutions, ready to use
  KRAF03087.3060
 :  
Buffer solutions, ready to use
Buffer solutions, ready to use

8,0, Phosphate, Colourless, 20 °C, Plastic bottle, 1 L

 :  
 
41,30 €
   
1 * 1 L
 
  • NIST traceable

 

 

 

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Phosphate
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • Colour:
    Colourless
  • pH value:
    8,0

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.