Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Buffer solutions, ready to use

5,0, Citrate, Colourless, 20 °C, Plastic bottle, 500 mL
Catalog # KRAF03084.2000
Supplier:   Bernd Kraft
Buffer solutions, ready to use
Catalog # KRAF03084.2000
Supplier:  Bernd Kraft
CAS Number:  
Buffer solutions, ready to use
Buffer solutions, ready to use

5,0, Citrate, Colourless, 20 °C, Plastic bottle, 500 mL

CAS Number:  
Price
24,70 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • NIST traceable

 

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Citrate
  • Pk:
    500 mL
  • Reference temperature:
    20 °C
  • Colour:
    Colourless
  • pH value:
    5,0

Specifications