Water
Plastic bottle, 5 L
KRAF01870.5000
: Hydrogen oxide
- :7732-18-5
- :H₂O
- :MFCD00011332
- :18.02 g/mol
- :13,10098
- :100 °C (1013 hPa)
- :0 °C
- :1 g/cm³ (4 °C)
- :Ambient
- :231-791-2
- Pack type:Plastic bottle
- Pk:5 L
- Cat. No.:KRAF01870.5000
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Environmentally Preferable:Safer Human and Environmental Health



