ContactUsLinkComponent
Palladium-magnesium nitrate matrix modifier for graphite furnace AAS 1,5 g Pd2+ and 1.00 g Mg(NO3)2 /l in nitric acid 2 mol/l Conten t: 0,5 l 1 * 500 mL
Catalog # KRAF03677.2000
Supplier:  Bernd Kraft
Palladium-magnesium nitrate matrix modifier for graphite furnace AAS 1,5 g Pd2+ and 1.00 g Mg(NO3)2 /l in nitric acid 2 mol/l Conten t: 0,5 l 1 * 500 mL
Catalog # KRAF03677.2000
Supplier:  Bernd Kraft
Frequently Bought Together