Order Entry
Germany
Orders LinkContactUsLinkComponent
D-(+)-Biotin ≥98%

D-(+)-Biotin ≥98%

Glass bottle for solids, 5 g
Catalog # A14207.06
D-(+)-Biotin ≥98%
Catalog # A14207.06
CAS Number:  58-85-5
D-(+)-Biotin ≥98%
D-(+)-Biotin ≥98%

Glass bottle for solids, 5 g

CAS Number:  58-85-5
Price
237,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 98+% CAS No.: 58-85-5 Molecular Formula: C10H16N2O3S Formula Weight: 244.32 Storage Temperature: 2°C to 8°C Melting Point: 224-226° Beilstein Reference: 86838 Merck Reference: 14,1231 Optical Rotation: +91° (c=1 in 0.1N NaOH) MDL No.: MFCD00005541

Essential vitamin that is important for amino acid and energy metabolism

Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Merck Index:14,1231
  • Melting Pt:224…226 °C
  • Density:1.41 g/cm³ (20 °C)
  • Storage Temperature:Refrigerator
  • EINECS:200-399-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications