Order Entry
Germany
Orders LinkContactUsLinkComponent
Sample cups for analysers

Sample cups for analysers

For Technicon analysers, with PE cap, 2,00 ml, 24,90 mm, 13,75 mm, Bag of 1000, 10 bags/case
Catalog # DELT910022
Supplier:   DELTALAB
Sample cups for analysers
Catalog # DELT910022
Supplier:  DELTALAB
Sample cups for analysers

For Technicon analysers, with PE cap, 2,00 ml, 24,90 mm, 13,75 mm, Bag of 1000, 10 bags/case

Price
422,00 €
Log in for availability
1 * 10.000 ST
Product Details & Documents

Cups made of PS.

Specifications
  • Ext.Ø:
    13,75 mm
  • Packed:
    Bag of 1000, 10 bags/case
  • Capacity:
    2,00 ml
  • Description:
    For Technicon analysers, with PE cap
  • Height:
    24,90 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.