Orders LinkContactUsLinkComponent
Inlet liners
Inlet liners
Inlet liner non-stick O-ring
Catalog # 548-0707
Supplier:   Agilent
Inlet liners
Catalog # 548-0707
Supplier:  Agilent
Inlet liners
Inlet liners
Inlet liner non-stick O-ring
Price
24,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
  • Consistent high level of inertness
Specifications
  • Description:
    Inlet liner non-stick O-ring
  • Seq:
    56